missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 414090010

 View more versions of this product

Catalog No. AC414090010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-2-butene
-134.0°C
0.6600g/mL
−45°C
Balance mainly 2-methyl-1-butene (GC)
C5H10
CH3CH=C(CH3)2
1 L
0.66
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CC=C(C)C
70.14
CHEBI:77916
90%
513-35-9
Colorless to Yellow
35.0°C to 38.0°C
Authentic
Glass bottle
1.3860 to 1.3880
MFCD00009276
01,211
15,601
BKOOMYPCSUNDGP-UHFFFAOYSA-N
2-methylbut-2-ene
10553
70.13
Liquid

Chemical Identifiers

513-35-9
70.14
BKOOMYPCSUNDGP-UHFFFAOYSA-N
10553
2-methylbut-2-ene
C5H10
MFCD00009276
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CHEBI:77916
CC=C(C)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected o

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a well-ventilated place.
Keep cool.
Keep away from heat/sparks/open flame

GHS Signal Word: Danger

EINECSNumber : 208-156-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only