missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene, Thermo Scientific Chemicals

$238.65 - $788.17

Chemical Identifiers

CAS 513-35-9
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009276
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
PubChem CID 10553
ChEBI CHEBI:77916
IUPAC Name 2-methylbut-2-ene
SMILES CC=C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414092500
SDS View Documents
Thermo Scientific Chemicals
414092500
250 mL Glass bottle
Each for $238.65
 
AC414090010
SDS View Documents
Thermo Scientific Chemicals
414090010
1 L Glass bottle
Each for $788.17
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-35-9
70.14
BKOOMYPCSUNDGP-UHFFFAOYSA-N
10553
2-methylbut-2-ene
C5H10
MFCD00009276
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CHEBI:77916
CC=C(C)C

Specifications

513-35-9
Colorless to Yellow
35.0°C to 38.0°C
Authentic
Glass bottle
1.3860 to 1.3880
MFCD00009276
01,211
15,601
BKOOMYPCSUNDGP-UHFFFAOYSA-N
2-methylbut-2-ene
10553
70.13
Liquid
-134.0°C
0.6600g/mL
−45°C
Balance mainly 2-methyl-1-butene (GC)
C5H10
CH3CH=C(CH3)2
250 mL
0.66
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CC=C(C)C
70.14
CHEBI:77916
90%
2-Methyl-2-butene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected o

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a well-ventilated place.
Keep cool.
Keep away from heat/sparks/open flame

GHS Signal Word: Danger

EINECSNumber : 208-156-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only