Learn More
2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene
CAS: 513-35-9 | C5H10 | 70.14 g/mol
Supplier: Thermo Scientific Chemicals 414092500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methyl-2-butene | |
| -134.0°C | |
| 0.6600g/mL | |
| −45°C | |
| Balance mainly 2-methyl-1-butene (GC) | |
| C5H10 | |
| CH3CH=C(CH3)2 | |
| 250 mL | |
| 0.66 | |
| 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2 | |
| CC=C(C)C | |
| 70.14 | |
| CHEBI:77916 | |
| 90% |
| 513-35-9 | |
| Colorless to Yellow | |
| 35.0°C to 38.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3860 to 1.3880 | |
| MFCD00009276 | |
| 01,211 | |
| 15,601 | |
| BKOOMYPCSUNDGP-UHFFFAOYSA-N | |
| 2-methylbut-2-ene | |
| 10553 | |
| 70.13 | |
| Liquid |
Chemical Identifiers
| 513-35-9 | |
| 70.14 | |
| BKOOMYPCSUNDGP-UHFFFAOYSA-N | |
| 10553 | |
| 2-methylbut-2-ene |
| C5H10 | |
| MFCD00009276 | |
| 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2 | |
| CHEBI:77916 | |
| CC=C(C)C |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected o
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a well-ventilated place.
Keep cool.
Keep away from heat/sparks/open flame
GHS Signal Word: Danger
EINECSNumber : 208-156-3
RUO – Research Use Only