missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-2H-indazole-3-carbaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC44404DE
Specifications
2-Methyl-2H-indazole-3-carbaldehyde | |
97% | |
C9H8N2O | |
5g | |
SMWLSZQHLMZHHV-UHFFFAOYSA-N | |
2-methylindazole-3-carbaldehyde | |
818117 | |
97% |
34252-54-5 | |
Amber Glass Bottle | |
MFCD00839530 | |
2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl | |
CN1N=C2C=CC=CC2=C1C=O | |
160.18 | |
160.18 |
Chemical Identifiers
34252-54-5 | |
160.18 | |
SMWLSZQHLMZHHV-UHFFFAOYSA-N | |
818117 | |
CN1N=C2C=CC=CC2=C1C=O |
C9H8N2O | |
MFCD00839530 | |
2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl | |
2-methylindazole-3-carbaldehyde |