missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-2H-indazole-3-carbaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 34252-54-5 |
---|---|
Molecular Formula | C9H8N2O |
Molecular Weight (g/mol) | 160.18 |
MDL Number | MFCD00839530 |
InChI Key | SMWLSZQHLMZHHV-UHFFFAOYSA-N |
Synonym | 2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl |
PubChem CID | 818117 |
IUPAC Name | 2-methylindazole-3-carbaldehyde |
SMILES | CN1N=C2C=CC=CC2=C1C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC44404CB
|
Thermo Scientific Chemicals
CC44404CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC44404DA
|
Thermo Scientific Chemicals
CC44404DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC44404DE
|
Thermo Scientific Chemicals
CC44404DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
34252-54-5 | |
160.18 | |
SMWLSZQHLMZHHV-UHFFFAOYSA-N | |
818117 | |
CN1N=C2C=CC=CC2=C1C=O |
C9H8N2O | |
MFCD00839530 | |
2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl | |
2-methylindazole-3-carbaldehyde |
Specifications
34252-54-5 | |
Amber Glass Bottle | |
MFCD00839530 | |
2-methyl-2h-indazole-3-carbaldehyde, 2-methyl-3-formylindazole, 2h-indazole-3-carboxaldehyde,2-methyl | |
CN1N=C2C=CC=CC2=C1C=O | |
160.18 | |
160.18 | |
2-Methyl-2H-indazole-3-carbaldehyde |
97% | |
C9H8N2O | |
250mg | |
SMWLSZQHLMZHHV-UHFFFAOYSA-N | |
2-methylindazole-3-carbaldehyde | |
818117 | |
97% |