Learn More
2-Methyl-3,5-dinitrobenzoic acid, 97+%, Thermo Scientific Chemicals
$48.77 - $299.49
Chemical Identifiers
CAS | 28169-46-2 |
---|---|
Molecular Formula | C8H6N2O6 |
Molecular Weight (g/mol) | 226.14 |
MDL Number | MFCD00007161 |
InChI Key | CDVNZMKTJIBBBV-UHFFFAOYSA-N |
Synonym | 2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro |
PubChem CID | 98824 |
IUPAC Name | 2-methyl-3,5-dinitrobenzoic acid |
SMILES | CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1455514
|
Thermo Scientific Chemicals
A1455514 |
25 g |
Each for $48.77
|
|
|||||
AAA1455522
|
Thermo Scientific Chemicals
A1455522 |
100 g |
Each for $93.88
|
|
|||||
AAA1455536
|
Thermo Scientific Chemicals
A1455536 |
500 g |
Each for $299.49
|
|
|||||
Description
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
28169-46-2 | |
226.14 | |
CDVNZMKTJIBBBV-UHFFFAOYSA-N | |
98824 | |
CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O |
C8H6N2O6 | |
MFCD00007161 | |
2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro | |
2-methyl-3,5-dinitrobenzoic acid |
Specifications
28169-46-2 | |
C8H6N2O6 | |
25 g | |
2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro | |
CDVNZMKTJIBBBV-UHFFFAOYSA-N | |
2-methyl-3,5-dinitrobenzoic acid | |
98824 | |
98% |
203°C to 207°C | |
MFCD00007161 | |
2622906 | |
Soluble in water. | |
CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O | |
226.14 | |
226.15 | |
2-Methyl-3,5-dinitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 248-880-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only