Learn More
2-Methyl-5-nitroaniline, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1772636
Description
It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.
Solubility
Insoluble in water. (<0.1 g/100 mL at 19°C). Solubility in methanol, almost transparency
Notes
C.I. 37105. Sensitive to moisture and light. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
Specifications
2-Methyl-5-nitroaniline | |
105°C to 108°C | |
C7H8N2O2 | |
500 g | |
879021 | |
Insoluble in water. (<0.1g/100 mL at 19°C). Solubility in methanol,almost transparency | |
CC1=CC=C(C=C1N)[N+]([O-])=O | |
152.15 | |
CHEBI:66891 | |
≥98% |
99-55-8 | |
Characteristic | |
MFCD00007741 | |
UN2660 | |
2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp | |
DSBIJCMXAIKKKI-UHFFFAOYSA-N | |
2-methyl-5-nitroaniline | |
7444 | |
152.15 |
Chemical Identifiers
99-55-8 | |
152.15 | |
DSBIJCMXAIKKKI-UHFFFAOYSA-N | |
7444 | |
2-methyl-5-nitroaniline |
C7H8N2O2 | |
MFCD00007741 | |
2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp | |
CHEBI:66891 | |
CC1=CC=C(C=C1N)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
H301+H311+H331-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
EINECSNumber : 202-765-8
RTECSNumber : XU8225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only