missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-5-nitroaniline, 98+%, Thermo Scientific Chemicals

$66.53 - $1009.56

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007741
InChI Key DSBIJCMXAIKKKI-UHFFFAOYSA-N
Synonym 2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
PubChem CID 7444
ChEBI CHEBI:66891
IUPAC Name 2-methyl-5-nitroaniline
SMILES CC1=CC=C(C=C1N)[N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1772622
View Documents
Thermo Scientific Chemicals
A1772622
100 g
Each for $66.53
 
AAA1772636
View Documents
Thermo Scientific Chemicals
A1772636
500 g
Each for $291.60
 
AAA177260E
View Documents
Thermo Scientific Chemicals
A177260E
2500 g
Each for $1,009.56
 
Description

Description

It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in organic synthesis. 5-Nitro-o-toluidine finds application as an intermediate in the synthesis of Pigment Red 17 (C.I. 12 390) and Pigment Red 22 (C.I. 12 315). It is also employed in conjunction with certain C.I. coupling components. It also serves as a precursor for a wide assortment of azo dyes of various red, yellow, orange, violet, and brown hues. It can also be employed as analytical standard for the surfactant mediated transformations (-254nm) of 2,4-dinitrotoluene.

Solubility
Insoluble in water. (<0.1 g/100 mL at 19°C). Solubility in methanol, almost transparency

Notes
C.I. 37105. Sensitive to moisture and light. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
Specifications

Chemical Identifiers

99-55-8
152.15
DSBIJCMXAIKKKI-UHFFFAOYSA-N
7444
2-methyl-5-nitroaniline
C7H8N2O2
MFCD00007741
2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
CHEBI:66891
CC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

99-55-8
Characteristic
MFCD00007741
UN2660
2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
DSBIJCMXAIKKKI-UHFFFAOYSA-N
2-methyl-5-nitroaniline
7444
152.15
2-Methyl-5-nitroaniline
105°C to 108°C
C7H8N2O2
100 g
879021
Insoluble in water. (<0.1g/100 mL at 19°C). Solubility in methanol,almost transparency
CC1=CC=C(C=C1N)[N+]([O-])=O
152.15
CHEBI:66891
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c

H301+H311+H331-H351

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)

EINECSNumber : 202-765-8

RTECSNumber : XU8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only