2-Methylbenzoic Anhydride 98.0+%, TCI America™
Supplier:  TCI America M29975G
 View more versions of this product
Catalog No. M29975G
Explore available promotions
Description
Description
Specifications
Specifications
2-Methylbenzoic Anhydride
39°C
221°C
MFCD00048077
YLBSXJWDERHYFY-UHFFFAOYSA-N
2-methylbenzoyl 2-methylbenzoate
136405
≥98.0% (GC,T)
607-86-3
White
C16H14O3
5 g
CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C
254.29
254.29
Crystalline Powder
Chemical Identifiers
607-86-3
254.29
YLBSXJWDERHYFY-UHFFFAOYSA-N
2-methylbenzoyl 2-methylbenzoate
C16H14O3
MFCD00048077
136405
CC1=CC=CC=C1C(=O)OC(=O)C1=CC=CC=C1C
Videos
Safety and Handling
Safety and Handling
TSCA : No