missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclohexane-1,3-dione, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1121806

 View more versions of this product

Catalog No. AAA1121806


Explore available promotions

Description

Description

2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in methanol ( 0.1 g/mL). Insoluble in water.

Notes
Hygroscopic. Store at -20°C. Incompatible with oxidizing agents and moisture.
Specifications

Specifications

2-Methylcyclohexane-1,3-dione
204°C to 208°C
MFCD00001587
1281598
2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g
VSGJHHIAMHUZKF-UHFFFAOYSA-N
2-methylcyclohexane-1,3-dione
70945
≥98%
1193-55-1
C7H10O2
5 g
Hygroscopic
Soluble in methanol ( 0.1g/mL). Insoluble in water.
CC1C(=O)CCCC1=O
126.16
126.16

Chemical Identifiers

1193-55-1
126.16
VSGJHHIAMHUZKF-UHFFFAOYSA-N
70945
CC1C(=O)CCCC1=O
C7H10O2
MFCD00001587
2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g
2-methylcyclohexane-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-773-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only