Learn More
2-Methylcyclohexane-1,3-dione, 98+%, Thermo Scientific Chemicals
$78.52 - $670.21
Chemical Identifiers
CAS | 1193-55-1 |
---|---|
Molecular Formula | C7H10O2 |
Molecular Weight (g/mol) | 126.16 |
MDL Number | MFCD00001587 |
InChI Key | VSGJHHIAMHUZKF-UHFFFAOYSA-N |
Synonym | 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g |
PubChem CID | 70945 |
IUPAC Name | 2-methylcyclohexane-1,3-dione |
SMILES | CC1C(=O)CCCC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1121806
|
Thermo Scientific Chemicals
A1121806 |
5 g |
Each for $78.52
|
|
||||||
AAA1121814
|
Thermo Scientific Chemicals
A1121814 |
25 g |
Each for $226.45
|
|
||||||
AAA1121822
|
Thermo Scientific Chemicals
A1121822 |
100 g |
Each for $670.21
|
|
||||||
Description
2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in methanol ( 0.1 g/mL). Insoluble in water.
Notes
Hygroscopic. Store at -20°C. Incompatible with oxidizing agents and moisture.
Chemical Identifiers
1193-55-1 | |
126.16 | |
VSGJHHIAMHUZKF-UHFFFAOYSA-N | |
70945 | |
CC1C(=O)CCCC1=O |
C7H10O2 | |
MFCD00001587 | |
2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g | |
2-methylcyclohexane-1,3-dione |
Specifications
1193-55-1 | |
C7H10O2 | |
5 g | |
Hygroscopic | |
Soluble in methanol ( 0.1g/mL). Insoluble in water. | |
CC1C(=O)CCCC1=O | |
126.16 | |
126.16 | |
2-Methylcyclohexane-1,3-dione |
204°C to 208°C | |
MFCD00001587 | |
1281598 | |
2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g | |
VSGJHHIAMHUZKF-UHFFFAOYSA-N | |
2-methylcyclohexane-1,3-dione | |
70945 | |
≥98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-773-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only