Learn More
2-Methylcyclohexane-1,3-dione, 98+%
CAS: 1193-55-1 | C7H10O2 | 126.16 g/mol
Supplier: Thermo Scientific Chemicals A1121814
Description
2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in methanol ( 0.1 g/mL). Insoluble in water.
Notes
Hygroscopic. Store at -20°C. Incompatible with oxidizing agents and moisture.
Specifications
| 2-Methylcyclohexane-1,3-dione | |
| 204°C to 208°C | |
| MFCD00001587 | |
| 1281598 | |
| 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g | |
| VSGJHHIAMHUZKF-UHFFFAOYSA-N | |
| 2-methylcyclohexane-1,3-dione | |
| 70945 | |
| ≥98% |
| 1193-55-1 | |
| C7H10O2 | |
| 25 g | |
| Hygroscopic | |
| Soluble in methanol ( 0.1g/mL). Insoluble in water. | |
| CC1C(=O)CCCC1=O | |
| 126.16 | |
| 126.16 |
Chemical Identifiers
| 1193-55-1 | |
| 126.16 | |
| VSGJHHIAMHUZKF-UHFFFAOYSA-N | |
| 70945 | |
| CC1C(=O)CCCC1=O |
| C7H10O2 | |
| MFCD00001587 | |
| 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g | |
| 2-methylcyclohexane-1,3-dione |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-773-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only