missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylcyclopentanone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 247660050

 View more versions of this product

Catalog No. AC247660050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylcyclopentanone
98.5
Colorless to Yellow
139°C to 140°C
Authentic
Glass bottle
1.4340 to 1.436
5 g
2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one
ZIXLDMFVRPABBX-UHFFFAOYNA-N
2-methylcyclopentan-1-one
14265
99%
1120-72-5
100.0
0.9100g/mL
26°C
99%
C6H10O
MFCD00001414
0.91
Solubility in water: soluble. Other solubilities: very soluble in ethanol,ether and acetone
CC1CCCC1=O
98.15
98.14
Liquid

Chemical Identifiers

1120-72-5
98.15
ZIXLDMFVRPABBX-UHFFFAOYNA-N
14265
CC1CCCC1=O
C6H10O
MFCD00001414
2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one
2-methylcyclopentan-1-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-318-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only