missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylimidazole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 127005000

 View more versions of this product

Catalog No. AC127005000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylimidazole
693-98-1
100.0
White to Yellow
155°C
98.5% min. (GC)
C4H6N2
500 g
2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
LXBGSDVWAMZHDD-UHFFFAOYSA-N
2-methyl-1H-imidazole
12749
99%
99%
98.5
142.0°C to 145.0°C
270.0°C
Authentic
Plastic bottle
MFCD00005190
23, 65
Solubility in water: 267g/L (20°C)
CC1=NC=CN1
82.11
82.1
Crystalline Powder

Chemical Identifiers

693-98-1
82.11
LXBGSDVWAMZHDD-UHFFFAOYSA-N
12749
CC1=NC=CN1
C4H6N2
MFCD00005190
2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
2-methyl-1H-imidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Suspected of causing cancer.
May damage fertility or the unborn child.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 211-765-7

RTECSNumber : NI7175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only