missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole, 99%

CAS: 693-98-1 | C4H6N2 | 82.11 g/mol

$66.34 - $335.13

Chemical Identifiers

CAS 693-98-1
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005190
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
Synonym 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
PubChem CID 12749
IUPAC Name 2-methyl-1H-imidazole
SMILES CC1=NC=CN1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127001000
SDS View Documents
Thermo Scientific Chemicals
127001000
100 g Plastic bottle
Each for $66.34
Only null left
 
AC127005000
SDS View Documents
Thermo Scientific Chemicals
127005000
500 g Plastic bottle
Each for $177.53
Only null left
 
AC127000010
SDS View Documents
Thermo Scientific Chemicals
127000010
1 kg Plastic bottle
Each for $335.13
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

693-98-1
82.11
LXBGSDVWAMZHDD-UHFFFAOYSA-N
12749
CC1=NC=CN1
C4H6N2
MFCD00005190
2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
2-methyl-1H-imidazole

Specifications

693-98-1
100.0
White to Yellow
155°C
98.5% min. (GC)
C4H6N2
100 g
2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
LXBGSDVWAMZHDD-UHFFFAOYSA-N
2-methyl-1H-imidazole
12749
99%
2-Methylimidazole, 99%
98.5
142.0°C to 145.0°C
270.0°C
Authentic
Plastic bottle
MFCD00005190
23, 65
Solubility in water: 267g/L (20°C)
CC1=NC=CN1
82.11
82.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Suspected of causing cancer.
May damage fertility or the unborn child.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 211-765-7

RTECSNumber : NI7175000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only