missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 127341001

 View more versions of this product

Catalog No. AC127341001


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylpentane
107-83-5
100.0
Colorless
60.0°C
Authentic
Glass bottle
1.3700 to 1.3720
MFCD00009406
01, 148
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
AFABGHUZZDYHJO-UHFFFAOYSA-N
2-methylpentane
7892
99+%
Liquid
Pure
99.0
-154.0°C
0.6600g/mL
−23°C
99% min. (GC)
C6H14
CH3CH2CH2CH(CH3)2
100 mL
0.66
Solubility in water: insoluble. Other solubilities: soluble in ethanol
CCCC(C)C
86.18
86.18
Pure

Chemical Identifiers

107-83-5
86.18
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
CCCC(C)C
C6H14
MFCD00009406
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
2-methylpentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and wa

GHS Signal Word: Danger

EINECSNumber : 203-523-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only