Learn More
2-Methylpentane, 99+%, pure, Thermo Scientific Chemicals
$179.12 - $2,624.46
Chemical Identifiers
CAS | 107-83-5 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD00009406 |
InChI Key | AFABGHUZZDYHJO-UHFFFAOYSA-N |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
PubChem CID | 7892 |
IUPAC Name | 2-methylpentane |
SMILES | CCCC(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC127341001
|
Thermo Scientific Chemicals
127341001 |
100 mL | Glass bottle |
N/A
|
|
||||
AC127340010
|
Thermo Scientific Chemicals
127340010 |
1 L | Glass bottle |
N/A
|
|
||||
AC127340025
|
Thermo Scientific Chemicals
127340025 |
2.5 L | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-83-5 | |
86.18 | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
7892 | |
CCCC(C)C |
C6H14 | |
MFCD00009406 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
2-methylpentane |
Specifications
107-83-5 | |
100.0 | |
Colorless | |
60.0°C | |
Authentic | |
Glass bottle | |
1.3700 to 1.3720 | |
MFCD00009406 | |
01, 148 | |
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane | |
AFABGHUZZDYHJO-UHFFFAOYSA-N | |
2-methylpentane | |
7892 | |
99+% | |
Liquid |
99.0 | |
-154.0°C | |
0.6600g/mL | |
−23°C | |
99% min. (GC) | |
C6H14 | |
CH3CH2CH2CH(CH3)2 | |
100 mL | |
0.66 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol | |
CCCC(C)C | |
86.18 | |
86.18 | |
Pure | |
2-Methylpentane, Pure |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and wa
GHS Signal Word: Danger
EINECSNumber : 203-523-4
RUO – Research Use Only