missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylpentyl Acetate 98.0+%, TCI America™
Supplier: TCI America A107925ML
Specifications
2-Methylpentyl Acetate | |
Colorless | |
C8H16O2 | |
25 mL | |
Acetic Acid 2-Methylpentyl Ester | |
CCCC(C)COC(C)=O | |
144.21 | |
144.21 | |
Liquid |
7789-99-3 | |
160°C | |
MFCD00060103 | |
3272 | |
UZTVUHTZGGZFCI-UHFFFAOYNA-N | |
2-methylpentyl acetate | |
24625 | |
≥98.0% (GC) |
Chemical Identifiers
7789-99-3 | |
144.21 | |
UZTVUHTZGGZFCI-UHFFFAOYNA-N | |
24625 | |
CCCC(C)COC(C)=O |
C8H16O2 | |
MFCD00060103 | |
Acetic Acid 2-Methylpentyl Ester | |
2-methylpentyl acetate |
Safety and Handling
EINECSNumber : (2)-0734
RTECSNumber : AI9300000
TSCA : Yes