missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylpentyl Acetate 98.0+%, TCI America™
$59.73 - $574.02
Chemical Identifiers
CAS | 7789-99-3 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.21 |
MDL Number | MFCD00060103 |
InChI Key | UZTVUHTZGGZFCI-UHFFFAOYNA-N |
Synonym | Acetic Acid 2-Methylpentyl Ester |
PubChem CID | 24625 |
IUPAC Name | 2-methylpentyl acetate |
SMILES | CCCC(C)COC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A107925ML
|
TCI America
A107925ML |
25 mL |
Each for $59.73
|
|
|||||
A1079500ML
|
TCI America
A1079500ML |
500 mL |
Each for $574.02
|
|
|||||
Chemical Identifiers
7789-99-3 | |
144.21 | |
UZTVUHTZGGZFCI-UHFFFAOYNA-N | |
24625 | |
CCCC(C)COC(C)=O |
C8H16O2 | |
MFCD00060103 | |
Acetic Acid 2-Methylpentyl Ester | |
2-methylpentyl acetate |
Specifications
7789-99-3 | |
160°C | |
MFCD00060103 | |
3272 | |
UZTVUHTZGGZFCI-UHFFFAOYNA-N | |
2-methylpentyl acetate | |
24625 | |
≥98.0% (GC) | |
2-Methylpentyl Acetate |
Colorless | |
C8H16O2 | |
25 mL | |
Acetic Acid 2-Methylpentyl Ester | |
CCCC(C)COC(C)=O | |
144.21 | |
144.21 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0734
RTECSNumber : AI9300000
TSCA : Yes