missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-2-Methylpiperazine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2440514

 View more versions of this product

Catalog No. AAB2440514

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+/-)-2-Methylpiperazine
62°C to 66°C
65°C (149°F)
C5H12N2
25 g
383566
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
CC1CNCCN1
100.165
100.17
109-07-9
155°C to 156°C
Amine-like
MFCD00005954
UN2925
Air sensitive; Hygroscopic
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
2-methylpiperazine
66057
98%

Chemical Identifiers

109-07-9
100.165
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
66057
CC1CNCCN1
C5H12N2
MFCD00005954
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
2-methylpiperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 203-644-2

RTECSNumber : TM1225020

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only