missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-2-Methylpiperazine, 98%, Thermo Scientific Chemicals

$81.63 - $219.14

Chemical Identifiers

CAS 109-07-9
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005954
InChI Key JOMNTHCQHJPVAZ-UHFFFAOYSA-N
Synonym piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
PubChem CID 66057
IUPAC Name 2-methylpiperazine
SMILES CC1CNCCN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2440514
View Documents
Thermo Scientific Chemicals
B2440514
25 g
Each for $81.63
 
AAB2440522
View Documents
Thermo Scientific Chemicals
B2440522
100 g
Each for $219.14
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-07-9
100.165
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
66057
CC1CNCCN1
C5H12N2
MFCD00005954
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
2-methylpiperazine

Specifications

109-07-9
155°C to 156°C
Amine-like
MFCD00005954
UN2925
Air sensitive; Hygroscopic
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
2-methylpiperazine
66057
98%
62°C to 66°C
65°C (149°F)
C5H12N2
25 g
383566
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
CC1CNCCN1
100.165
100.17
(+/-)-2-Methylpiperazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 203-644-2

RTECSNumber : TM1225020

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only