missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylpiperazine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 127471000

 View more versions of this product

Catalog No. AC127471000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methylpiperazine
61.0°C to 66.0°C
154.0°C to 156.0°C
Authentic
Glass bottle
100 g
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
2-methylpiperazine
66057
98%
109-07-9
White to Yellow
63°C
97.5% min. (GC)
C5H12N2
23,17
Solubility in water: 78% (25°C).
CC1CNCCN1
100.16
100.16
Crystalline Powder, Crystals or Chunks

Chemical Identifiers

109-07-9
100.16
piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
2-methylpiperazine
C5H12N2
JOMNTHCQHJPVAZ-UHFFFAOYSA-N
66057
CC1CNCCN1
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 203-644-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only