missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltryptamine 98.0+%, TCI America™
Supplier: TCI America M20555G
Specifications
| 2-Methyltryptamine | |
| 108°C | |
| C11H14N2 | |
| 5 g | |
| CPVSLHQIPGTMLH-UHFFFAOYSA-N | |
| 2-(2-methyl-1H-indol-3-yl)ethanamine | |
| 75949 | |
| ≥98.0% (GC,T) |
| 2731-06-8 | |
| 177°C | |
| MFCD00130185 | |
| 3-(2-Aminoethyl)-2-methylindole | |
| CC1=C(C2=CC=CC=C2N1)CCN | |
| 174.25 | |
| 174.25 | |
| Crystalline Powder |
Chemical Identifiers
| 2731-06-8 | |
| 174.25 | |
| CPVSLHQIPGTMLH-UHFFFAOYSA-N | |
| 75949 | |
| CC1=C(C2=CC=CC=C2N1)CCN |
| C11H14N2 | |
| MFCD00130185 | |
| 3-(2-Aminoethyl)-2-methylindole | |
| 2-(2-methyl-1H-indol-3-yl)ethanamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator