missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyltryptamine 98.0+%, TCI America™
$283.33 - $853.73
Chemical Identifiers
| CAS | 2731-06-8 |
|---|---|
| Molecular Formula | C11H14N2 |
| Molecular Weight (g/mol) | 174.25 |
| MDL Number | MFCD00130185 |
| InChI Key | CPVSLHQIPGTMLH-UHFFFAOYSA-N |
| Synonym | 3-(2-Aminoethyl)-2-methylindole |
| PubChem CID | 75949 |
| IUPAC Name | 2-(2-methyl-1H-indol-3-yl)ethanamine |
| SMILES | CC1=C(C2=CC=CC=C2N1)CCN |
Chemical Identifiers
| 2731-06-8 | |
| 174.25 | |
| CPVSLHQIPGTMLH-UHFFFAOYSA-N | |
| 75949 | |
| CC1=C(C2=CC=CC=C2N1)CCN |
| C11H14N2 | |
| MFCD00130185 | |
| 3-(2-Aminoethyl)-2-methylindole | |
| 2-(2-methyl-1H-indol-3-yl)ethanamine |
Specifications
| 2731-06-8 | |
| 177°C | |
| MFCD00130185 | |
| 3-(2-Aminoethyl)-2-methylindole | |
| CC1=C(C2=CC=CC=C2N1)CCN | |
| 174.25 | |
| 174.25 | |
| Crystalline Powder |
| 108°C | |
| C11H14N2 | |
| 1 g | |
| CPVSLHQIPGTMLH-UHFFFAOYSA-N | |
| 2-(2-methyl-1H-indol-3-yl)ethanamine | |
| 75949 | |
| ≥98.0% (GC,T) | |
| 2-Methyltryptamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator