missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 156972500

 View more versions of this product

Catalog No. AC156972500

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Naphthol
97.5
120.0°C to 124.0°C
285.0°C to 286.0°C
Authentic
Plastic bottle
C10H7OH
250 g
15,6469
Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol
C1=CC=C2C=C(C=CC2=C1)O
144.17
CHEBI:10432
98%
135-19-3
100.0
White to Yellow
153°C
97.5% min. (GC)
C10H8O
MFCD00004067
06,627
2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
JWAZRIHNYRIHIV-UHFFFAOYSA-N
naphthalen-2-ol
8663
144.17
Crystalline Powder or Crystals

Chemical Identifiers

135-19-3
144.17
JWAZRIHNYRIHIV-UHFFFAOYSA-N
8663
naphthalen-2-ol
C10H8O
MFCD00004067
2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
CHEBI:10432
C1=CC=C2C=C(C=CC2=C1)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 205-182-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only