Learn More
2-Naphthol, 98%
CAS: 135-19-3 | C10H8O | 144.17 g/mol
$60.23 - $60.23
Chemical Identifiers
| CAS | 135-19-3 |
|---|---|
| Molecular Formula | C10H8O |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00004067 |
| InChI Key | JWAZRIHNYRIHIV-UHFFFAOYSA-N |
| Synonym | 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol |
| PubChem CID | 8663 |
| ChEBI | CHEBI:10432 |
| IUPAC Name | naphthalen-2-ol |
| SMILES | C1=CC=C2C=C(C=CC2=C1)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC156972500
|
Thermo Scientific Chemicals
156972500 |
250 g | Plastic bottle |
Each for $60.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 135-19-3 | |
| 144.17 | |
| JWAZRIHNYRIHIV-UHFFFAOYSA-N | |
| 8663 | |
| naphthalen-2-ol |
| C10H8O | |
| MFCD00004067 | |
| 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol | |
| CHEBI:10432 | |
| C1=CC=C2C=C(C=CC2=C1)O |
Specifications
| 135-19-3 | |
| 100.0 | |
| White to Yellow | |
| 153°C | |
| 97.5% min. (GC) | |
| C10H8O | |
| MFCD00004067 | |
| 06,627 | |
| 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol | |
| JWAZRIHNYRIHIV-UHFFFAOYSA-N | |
| naphthalen-2-ol | |
| 8663 | |
| 144.17 | |
| Crystalline Powder or Crystals |
| 97.5 | |
| 120.0°C to 124.0°C | |
| 285.0°C to 286.0°C | |
| Authentic | |
| Plastic bottle | |
| C10H7OH | |
| 250 g | |
| 15,6469 | |
| Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol | |
| C1=CC=C2C=C(C=CC2=C1)O | |
| 144.17 | |
| CHEBI:10432 | |
| 98% | |
| 2-Naphthol |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 205-182-7
RUO – Research Use Only