missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1541236

 View more versions of this product

Catalog No. AAA1541236


Explore available promotions

Description

Description

2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.

Notes
Store in a cool place. Incompatible with oxidizing agents.
Specifications

Specifications

2-Naphthoxyacetic acid
155°C to 157°C
MFCD00004066
1074148
2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
RZCJYMOBWVJQGV-UHFFFAOYSA-N
2-naphthalen-2-yloxyacetic acid
8422
202.21
120-23-0
C12H10O3
500 g
14,6397
Soluble in alcohol,ether,acetic acid and dimethyl sulfoxide. Slightly soluble in water.
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
202.209
CHEBI:50397
97%

Chemical Identifiers

120-23-0
202.209
RZCJYMOBWVJQGV-UHFFFAOYSA-N
8422
2-naphthalen-2-yloxyacetic acid
C12H10O3
MFCD00004066
2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
CHEBI:50397
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 204-380-0

RTECSNumber : AI9659000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only