Learn More
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals
$66.38 - $192.55
Chemical Identifiers
CAS | 120-23-0 |
---|---|
Molecular Formula | C12H10O3 |
Molecular Weight (g/mol) | 202.209 |
MDL Number | MFCD00004066 |
InChI Key | RZCJYMOBWVJQGV-UHFFFAOYSA-N |
Synonym | 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal |
PubChem CID | 8422 |
ChEBI | CHEBI:50397 |
IUPAC Name | 2-naphthalen-2-yloxyacetic acid |
SMILES | C1=CC=C2C=C(C=CC2=C1)OCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1541222
|
Thermo Scientific Chemicals
A1541222 |
100 g |
Each for $66.38
|
|
||||||
AAA1541236
|
Thermo Scientific Chemicals
A1541236 |
500 g |
Each for $192.55
|
|
||||||
Description
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.
Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.
Notes
Store in a cool place. Incompatible with oxidizing agents.
Chemical Identifiers
120-23-0 | |
202.209 | |
RZCJYMOBWVJQGV-UHFFFAOYSA-N | |
8422 | |
2-naphthalen-2-yloxyacetic acid |
C12H10O3 | |
MFCD00004066 | |
2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal | |
CHEBI:50397 | |
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O |
Specifications
120-23-0 | |
C12H10O3 | |
100 g | |
14,6397 | |
Soluble in alcohol,ether,acetic acid and dimethyl sulfoxide. Slightly soluble in water. | |
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O | |
202.209 | |
CHEBI:50397 | |
97% |
155°C to 157°C | |
MFCD00004066 | |
1074148 | |
2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal | |
RZCJYMOBWVJQGV-UHFFFAOYSA-N | |
2-naphthalen-2-yloxyacetic acid | |
8422 | |
202.21 | |
2-Naphthoxyacetic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 204-380-0
RTECSNumber : AI9659000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only