missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04378704

 View more versions of this product

Catalog No. AA4378704


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Nitro-4- (trifluoromethyl)benzoic acid
C8H4F3NO4
2 g
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
235.118
235.12
Powder
320-94-5
MFCD00007142
2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
642096
98%

Chemical Identifiers

320-94-5
235.118
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
642096
C8H4F3NO4
MFCD00007142
2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only