missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitro-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

$96.96 - $259.32

Chemical Identifiers

CAS 320-94-5
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.118
MDL Number MFCD00007142
InChI Key MYSAXQPTXWKDPQ-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
PubChem CID 642096
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4378704
View Documents
Thermo Scientific Chemicals
04378704
2 g
Each for $96.96
 
AA4378709
View Documents
Thermo Scientific Chemicals
04378709
10 g
Each for $259.32
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

320-94-5
235.118
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
642096
C8H4F3NO4
MFCD00007142
2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O

Specifications

320-94-5
MFCD00007142
2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
642096
98%
2-Nitro-4- (trifluoromethyl)benzoic acid
C8H4F3NO4
2 g
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
235.118
235.12
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only