missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Nitrophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 128741000

 View more versions of this product

Catalog No. AC128741000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

2-Nitrophenol
88-75-5
100.0
Yellow
108°C
98.5% min. (GC)
C6H5NO3
MFCD00011688
1663
05,477
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
IQUPABOKLQSFBK-UHFFFAOYSA-N
2-nitrophenol
6947
CHEBI:16260
99%
99%
98.5
43.0°C to 47.0°C
214.0°C to 216.0°C
Authentic
Glass bottle
O2NC6H4OH
100 g
06, 213
15, 6707
Solubility in water: 2g/L (25°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene,carbon disulfide,alkali hydroxides,and toluene
OC1=CC=CC=C1[N+]([O-])=O
139.11
1.35 mPa.s (80°C)
139.11
Liquid

Chemical Identifiers

88-75-5
139.11
IQUPABOKLQSFBK-UHFFFAOYSA-N
6947
2-nitrophenol
C6H5NO3
MFCD00011688
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
CHEBI:16260
OC1=CC=CC=C1[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

GHS Signal Word: Warning

EINECSNumber : 201-857-5

RTECSNumber : SM2100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only