missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol, 99%

CAS: 88-75-5 | C6H5NO3 | 139.11 g/mol

$56.89 - $251.18

Chemical Identifiers

CAS 88-75-5
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011688
InChI Key IQUPABOKLQSFBK-UHFFFAOYSA-N
Synonym o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
PubChem CID 6947
ChEBI CHEBI:16260
IUPAC Name 2-nitrophenol
SMILES OC1=CC=CC=C1[N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128741000
SDS View Documents
Thermo Scientific Chemicals
128741000
100 g Glass bottle
Each for $56.89
Only null left
 
AC128745000
SDS View Documents
Thermo Scientific Chemicals
128745000
500 g Glass bottle
Each for $148.45
Only null left
 
AC128740010
SDS View Documents
Thermo Scientific Chemicals
128740010
1 kg Glass bottle
Each for $251.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

88-75-5
139.11
IQUPABOKLQSFBK-UHFFFAOYSA-N
6947
2-nitrophenol
C6H5NO3
MFCD00011688
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
CHEBI:16260
OC1=CC=CC=C1[N+]([O-])=O

Specifications

88-75-5
100.0
Yellow
108°C
98.5% min. (GC)
C6H5NO3
MFCD00011688
1663
05,477
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
IQUPABOKLQSFBK-UHFFFAOYSA-N
2-nitrophenol
6947
CHEBI:16260
99%
2-Nitrophenol, 99%
98.5
43.0°C to 47.0°C
214.0°C to 216.0°C
Authentic
Glass bottle
O2NC6H4OH
100 g
06, 213
15, 6707
Solubility in water: 2g/L (25°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene,carbon disulfide,alkali hydroxides,and toluene
OC1=CC=CC=C1[N+]([O-])=O
139.11
1.35 mPa.s (80°C)
139.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

GHS Signal Word: Warning

EINECSNumber : 201-857-5

RTECSNumber : SM2100000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only