Learn More
2-Octanone, 99+%, Thermo Scientific Chemicals
$99.51 - $154.86
Chemical Identifiers
CAS | 111-13-7 |
---|---|
MDL Number | MFCD00009540 |
InChI Key | ZPVFWPFBNIEHGJ-UHFFFAOYSA-N |
Synonym | 2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af |
PubChem CID | 8093 |
ChEBI | CHEBI:87434 |
IUPAC Name | octan-2-one |
SMILES | CCCCCCC(=O)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC129420250
|
Thermo Scientific Chemicals
129420250 |
25 mL | Glass bottle |
Each for $99.51
|
|
||||
AC129425000
|
Thermo Scientific Chemicals
129425000 |
500 mL | Glass bottle |
Each for $154.86
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-13-7 | |
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N | |
8093 | |
octan-2-one |
MFCD00009540 | |
2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af | |
CHEBI:87434 | |
CCCCCCC(=O)C |
Specifications
111-13-7 | |
100.0 | |
Colorless to Yellow | |
173°C | |
Authentic | |
Glass bottle | |
1.4140 to 1.416 | |
MFCD00009540 | |
01, 704 | |
15, 4747 | |
Solubility in water: 0.9g/L. Other solubilities: miscible with alcohol,ether,hydrocarbons and,esters | |
CCCCCCC(=O)C | |
128.21 | |
CHEBI:87434 | |
99+% | |
2-Octanone, 99+% |
99.0 | |
-16°C | |
0.8190g/mL | |
55°C | |
99% min. (GC) | |
C8H16O | |
CH3(CH2)5COCH3 | |
25 mL | |
0.819 | |
2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af | |
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N | |
octan-2-one | |
8093 | |
128.21 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-837-1
RTECSNumber : RH1484000
TSCA : TSCA
RUO – Research Use Only