missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Octene (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America O0042500ML
Specifications
2-Octene (cis- and trans- mixture) | |
Colorless | |
C8H16 | |
500 mL | |
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2 | |
CCCCCC=CC | |
112.216 | |
112.22 | |
Liquid |
111-67-1 | |
125°C | |
MFCD00009532 | |
3295 | |
ILPBINAXDRFYPL-HWKANZROSA-N | |
(E)-oct-2-ene | |
5364448 | |
≥98.0% (GC) |
Chemical Identifiers
111-67-1 | |
112.216 | |
ILPBINAXDRFYPL-HWKANZROSA-N | |
5364448 | |
CCCCCC=CC |
C8H16 | |
MFCD00009532 | |
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2 | |
(E)-oct-2-ene |
Safety and Handling
EINECSNumber : (2)-0024
TSCA : Yes