missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Octene (cis- and trans- mixture) 98.0+%, TCI America™
$119.44 - $920.75
Chemical Identifiers
CAS | 111-67-1 |
---|---|
Molecular Formula | C8H16 |
Molecular Weight (g/mol) | 112.216 |
MDL Number | MFCD00009532 |
InChI Key | ILPBINAXDRFYPL-HWKANZROSA-N |
Synonym | trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2 |
PubChem CID | 5364448 |
IUPAC Name | (E)-oct-2-ene |
SMILES | CCCCCC=CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
O004225ML
|
TCI America
O004225ML |
25 mL |
Each for $119.44
|
|
|||||
O0042500ML
|
TCI America
O0042500ML |
500 mL |
Each for $920.75
|
|
|||||
Chemical Identifiers
111-67-1 | |
112.216 | |
ILPBINAXDRFYPL-HWKANZROSA-N | |
5364448 | |
CCCCCC=CC |
C8H16 | |
MFCD00009532 | |
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2 | |
(E)-oct-2-ene |
Specifications
111-67-1 | |
125°C | |
MFCD00009532 | |
3295 | |
ILPBINAXDRFYPL-HWKANZROSA-N | |
(E)-oct-2-ene | |
5364448 | |
≥98.0% (GC) | |
2-Octene (cis- and trans- mixture) |
Colorless | |
C8H16 | |
25 mL | |
trans-2-octene, 2-octene, e-2-octene, e-oct-2-ene, 2-octene, e, 2-octene, 2e, unii-g13g1yr8yw, oct-2-ene, g13g1yr8yw, octene-2 | |
CCCCCC=CC | |
112.216 | |
112.22 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0024
TSCA : Yes