Learn More
2-Phenoxyethanol, 99%, Thermo Scientific Chemicals
2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C
$47.70 - $1361.55
Chemical Identifiers
CAS | 122-99-6 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00002857 |
InChI Key | QCDWFXQBSFUVSP-UHFFFAOYSA-N |
Synonym | phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane |
PubChem CID | 31236 |
ChEBI | CHEBI:64275 |
IUPAC Name | 2-phenoxyethanol |
SMILES | C1=CC=C(C=C1)OCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC130242500
|
Thermo Scientific Chemicals
130242500 |
250 mL | Glass bottle |
Each for $47.70
|
|
||||
AC130240010
|
Thermo Scientific Chemicals
130240010 |
1 L | Glass bottle |
Each for $107.58
|
|
||||
AC130240025
|
Thermo Scientific Chemicals
130240025 |
2.5 L | Glass bottle |
Each for $214.98
|
|
||||
AC130240051
|
Thermo Scientific Chemicals
130240051 |
5 L | Plastic drum |
Each for $442.73
|
|
||||
AC130240250
|
Thermo Scientific Chemicals
130240250 |
25 L | Metal drum |
Each for $1,361.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
122-99-6 | |
138.17 | |
QCDWFXQBSFUVSP-UHFFFAOYSA-N | |
31236 | |
2-phenoxyethanol |
C8H10O2 | |
MFCD00002857 | |
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane | |
CHEBI:64275 | |
C1=CC=C(C=C1)OCCO |
Specifications
122-99-6 | |
100.0 | |
Colorless | |
237.0°C | |
Authentic | |
Glass bottle | |
1.5360 to 1.5400 | |
MFCD00002857 | |
06,146 | |
15,7369 | |
Solubility in water: 30g/L (20°C). Other solubilities: freely soluble in alcohol and ether,miscible with acetone and ethyl acetate | |
C1=CC=C(C=C1)OCCO | |
138.17 | |
29 mPa.s (20°C) | |
138.17 | |
Liquid |
98.5 | |
11.0°C to 13.0°C | |
1.1000g/mL | |
130°C | |
98.5% min. (GC) | |
C8H10O2 | |
C6H5OCH2CH2OH | |
250 mL | |
1.1 | |
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane | |
QCDWFXQBSFUVSP-UHFFFAOYSA-N | |
2-phenoxyethanol | |
31236 | |
CHEBI:64275 | |
99% | |
2-Phenoxyethanol |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-589-7
RUO – Research Use Only