missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenyl-1,3-thiazole-4-carbaldehyde, ≥90%, Thermo Scientific™

Chemical Identifiers

CAS 20949-81-9
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.232
MDL Number MFCD02681934
InChI Key OLLKCCGWRITPOV-UHFFFAOYSA-N
Synonym 2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
PubChem CID 736524
IUPAC Name 2-phenyl-1,3-thiazole-4-carbaldehyde
SMILES C1=CC=C(C=C1)C2=NC(=CS2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC18304CB
SDS View Documents
Thermo Scientific Chemicals
CC18304CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18304DA
SDS View Documents
Thermo Scientific Chemicals
CC18304DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

20949-81-9
189.232
OLLKCCGWRITPOV-UHFFFAOYSA-N
736524
C1=CC=C(C=C1)C2=NC(=CS2)C=O
C10H7NOS
MFCD02681934
2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
2-phenyl-1,3-thiazole-4-carbaldehyde

Specifications

20949-81-9
100.0
Amber Glass Bottle
MFCD02681934
2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
C1=CC=C(C=C1)C2=NC(=CS2)C=O
189.232
189.24
2-Phenyl-1, 3-thiazole-4-carbaldehyde
94.5
95%
C10H7NOS
250mg
OLLKCCGWRITPOV-UHFFFAOYSA-N
2-phenyl-1,3-thiazole-4-carbaldehyde
736524
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'