missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol, 99%

CAS: 617-94-7 | C9H12O | 136.19 g/mol

$97.71 - $215.84

Identifiants chimiques

CAS 617-94-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004456
InChI Key BDCFWIDZNLCTMF-UHFFFAOYSA-N
Synonym 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
PubChem CID 12053
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)C1=CC=CC=C1
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC130830050
FDS Afficher les documents
Thermo Scientific Chemicals
130830050
5 g Glass bottle
chaque for $97.71
Il en reste null
 
AC130830250
FDS Afficher les documents
Thermo Scientific Chemicals
130830250
25 g Glass bottle
chaque for $215.84
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

617-94-7
136.19
BDCFWIDZNLCTMF-UHFFFAOYSA-N
12053
CC(C)(O)C1=CC=CC=C1
C9H12O
MFCD00004456
2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
2-phenylpropan-2-ol

Spécifications

617-94-7
100.0
Colorless to Yellow
88°C to 90°C (11.0 mmHg)
Authentic
Glass bottle
1.5190 to 1.522
MFCD00004456
06, 506
2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
BDCFWIDZNLCTMF-UHFFFAOYSA-N
2-phenylpropan-2-ol
12053
99%
2-Phenyl-2-propanol
98.5
32°C to 34°C
0.9730g/mL
88°C
98.5% min. (GC)
C9H12O
C6H5C(CH3)2OH
5 g
0.973
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene
CC(C)(O)C1=CC=CC=C1
136.19
136.19
Liquid After Melting
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-539-5

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only