missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Phenylthio)ethanol 97.0+%, TCI America™
$53.32 - $495.73
Chemical Identifiers
CAS | 699-12-7 |
---|---|
Molecular Formula | C8H10OS |
Molecular Weight (g/mol) | 154.23 |
MDL Number | MFCD00002905 |
InChI Key | KWWZHCSQVRVQGF-UHFFFAOYSA-N |
Synonym | 2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784 |
PubChem CID | 69685 |
IUPAC Name | 2-(phenylsulfanyl)ethan-1-ol |
SMILES | OCCSC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P073325ML
|
TCI America
P073325ML |
25 mL |
Each for $53.32
|
|
|||||
P0733500ML
|
TCI America
P0733500ML |
500 mL |
Each for $495.73
|
|
|||||
Chemical Identifiers
699-12-7 | |
154.23 | |
KWWZHCSQVRVQGF-UHFFFAOYSA-N | |
69685 | |
OCCSC1=CC=CC=C1 |
C8H10OS | |
MFCD00002905 | |
2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784 | |
2-(phenylsulfanyl)ethan-1-ol |
Specifications
699-12-7 | |
275°C | |
MFCD00002905 | |
2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784 | |
OCCSC1=CC=CC=C1 | |
154.23 | |
154.23 | |
Liquid |
Yellow | |
C8H10OS | |
25 mL | |
KWWZHCSQVRVQGF-UHFFFAOYSA-N | |
2-(phenylsulfanyl)ethan-1-ol | |
69685 | |
≥97.0% (GC) | |
2-(Phenylthio)ethanol |
Safety and Handling
TSCA : Yes