2-(Phenylthio)ethanol 97.0+%, TCI America™
Supplier:  TCI America P0733500ML
 View more versions of this product
Catalog No. P0733500ML
Explore available promotions
Description
Description
Specifications
Specifications
2-(Phenylthio)ethanol
Yellow
C8H10OS
500 mL
KWWZHCSQVRVQGF-UHFFFAOYSA-N
2-(phenylsulfanyl)ethan-1-ol
69685
≥97.0% (GC)
699-12-7
275°C
MFCD00002905
2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784
OCCSC1=CC=CC=C1
154.23
154.23
Liquid
Chemical Identifiers
699-12-7
154.23
KWWZHCSQVRVQGF-UHFFFAOYSA-N
69685
OCCSC1=CC=CC=C1
C8H10OS
MFCD00002905
2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784
2-(phenylsulfanyl)ethan-1-ol
Videos
Safety and Handling
Safety and Handling
TSCA : Yes