missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Picolyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 172120250

 View more versions of this product

Catalog No. AC172120250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Picolyl chloride hydrochloride
6959-47-3
100.0
Brown to Green-Yellow
98%
C6H6ClN·HCl
25 g
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-(chloromethyl)pyridine;hydrochloride
23392
164.04
Crystalline Mass
98%
97.5
119.0°C to 125.0°C
Authentic
Glass bottle
MFCD00012811
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
C1=CC=NC(=C1)CCl.Cl
164.04
CHEBI:76600
98%

Chemical Identifiers

6959-47-3
164.04
JPMRGPPMXHGKRO-UHFFFAOYSA-N
23392
2-(chloromethyl)pyridine;hydrochloride
C6H6ClN·HCl
MFCD00012811
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
CHEBI:76600
C1=CC=NC(=C1)CCl.Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 230-149-9

RTECSNumber : US6825000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only