Learn More
2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0243114
Description
2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to moisture.
Specifications
2-Propanesulfonyl chloride | |
-47°C | |
95°C (45 mmHg) | |
C3H7ClO2S | |
MFCD00007453 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CC(C)S(=O)(=O)Cl | |
142.597 | |
142.6 |
10147-37-2 | |
1.273 | |
86°C (186°F) | |
1.456 | |
25 g | |
1747497 | |
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride | |
DRINJBFRTLBHNF-UHFFFAOYSA-N | |
propane-2-sulfonyl chloride | |
82408 | |
97% |
Chemical Identifiers
10147-37-2 | |
142.597 | |
DRINJBFRTLBHNF-UHFFFAOYSA-N | |
82408 | |
CC(C)S(=O)(=O)Cl |
C3H7ClO2S | |
MFCD00007453 | |
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride | |
propane-2-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 233-415-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only