missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals

$85.10 - $255.67

Chemical Identifiers

CAS 10147-37-2
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007453
InChI Key DRINJBFRTLBHNF-UHFFFAOYSA-N
Synonym isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
PubChem CID 82408
IUPAC Name propane-2-sulfonyl chloride
SMILES CC(C)S(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0243106
View Documents
Thermo Scientific Chemicals
L0243106
5 g
Each for $85.10
 
AAL0243114
View Documents
Thermo Scientific Chemicals
L0243114
25 g
Each for $255.67
 
Description

Description

2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to moisture.
Specifications

Chemical Identifiers

10147-37-2
142.597
DRINJBFRTLBHNF-UHFFFAOYSA-N
82408
CC(C)S(=O)(=O)Cl
C3H7ClO2S
MFCD00007453
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
propane-2-sulfonyl chloride

Specifications

10147-37-2
1.273
86°C (186°F)
1.456
5 g
1747497
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
DRINJBFRTLBHNF-UHFFFAOYSA-N
propane-2-sulfonyl chloride
82408
97%
-47°C
95°C (45 mmHg)
C3H7ClO2S
MFCD00007453
UN3265
Moisture sensitive
Reacts with water.
CC(C)S(=O)(=O)Cl
142.597
142.6
2-Propanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 233-415-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only