missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Thiopheneethylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1092003

 View more versions of this product

Catalog No. AAA1092003


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Thiopheneethylamine
1.087
87°C (188°F)
1.551
1 g
106962
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
C1=CSC(=C1)CCN
127.205
127.21
30433-91-1
201°C
C6H9NS
MFCD00051495
UN2735
Air sensitive
HVLUYXIJZLDNIS-UHFFFAOYSA-N
2-thiophen-2-ylethanamine
116521
98%

Chemical Identifiers

30433-91-1
127.205
HVLUYXIJZLDNIS-UHFFFAOYSA-N
116521
C1=CSC(=C1)CCN
C6H9NS
MFCD00051495
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene
2-thiophen-2-ylethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H312-H314-H332-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 250-196-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only