Learn More
2-Thiopheneethylamine, 98%, Thermo Scientific Chemicals
$73.59 - $715.86
Chemical Identifiers
CAS | 30433-91-1 |
---|---|
Molecular Formula | C6H9NS |
Molecular Weight (g/mol) | 127.205 |
MDL Number | MFCD00051495 |
InChI Key | HVLUYXIJZLDNIS-UHFFFAOYSA-N |
Synonym | thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene |
PubChem CID | 116521 |
IUPAC Name | 2-thiophen-2-ylethanamine |
SMILES | C1=CSC(=C1)CCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1092003
|
Thermo Scientific Chemicals
A1092003 |
1 g |
Each for $73.59
|
|
||||||
AAA1092006
|
Thermo Scientific Chemicals
A1092006 |
5 g |
Each for $222.79
|
|
||||||
AAA1092014
|
Thermo Scientific Chemicals
A1092014 |
25 g |
Each for $715.86
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30433-91-1 | |
127.205 | |
HVLUYXIJZLDNIS-UHFFFAOYSA-N | |
116521 | |
C1=CSC(=C1)CCN |
C6H9NS | |
MFCD00051495 | |
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene | |
2-thiophen-2-ylethanamine |
Specifications
30433-91-1 | |
201°C | |
C6H9NS | |
MFCD00051495 | |
UN2735 | |
Air sensitive | |
HVLUYXIJZLDNIS-UHFFFAOYSA-N | |
2-thiophen-2-ylethanamine | |
116521 | |
98% |
1.087 | |
87°C (188°F) | |
1.551 | |
1 g | |
106962 | |
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene | |
C1=CSC(=C1)CCN | |
127.205 | |
127.21 | |
2-Thiopheneethylamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H312-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 250-196-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only