Learn More
2-Thiopheneethylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1092014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Thiopheneethylamine | |
1.087 | |
87°C (188°F) | |
1.551 | |
25 g | |
106962 | |
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene | |
C1=CSC(=C1)CCN | |
127.205 | |
127.21 |
30433-91-1 | |
201°C | |
C6H9NS | |
MFCD00051495 | |
UN2735 | |
Air sensitive | |
HVLUYXIJZLDNIS-UHFFFAOYSA-N | |
2-thiophen-2-ylethanamine | |
116521 | |
98% |
Chemical Identifiers
30433-91-1 | |
127.205 | |
HVLUYXIJZLDNIS-UHFFFAOYSA-N | |
116521 | |
C1=CSC(=C1)CCN |
C6H9NS | |
MFCD00051495 | |
thiophene-2-ethylamine, 2-thiopheneethylamine, 2-thiophen-2-yl ethanamine, thiopheneethanamine, 2-2-thienyl ethylamine, 2-thien-2-ylethanamine, 2-thiopheneethanamine, thiopheneethylamine, 2-thiophen-2-yl ethan-1-amine, 2-2-aminoethyl thiophene | |
2-thiophen-2-ylethanamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H312-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 250-196-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only