missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Thiophenemethanol, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 138860250

 View more versions of this product

Catalog No. AC138860250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Thiophenemethanol
99.0
0.9000g/mL
>110°C
99% min. (GC)
C5H6OS
MFCD00005454
17, 113
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
ZPHGMBGIFODUMF-UHFFFAOYSA-N
(thiophen-2-yl)methanol
69467
99+%
636-72-6
100.0
206.0°C to 207.0°C (761.0mmHg)
Authentic
Glass bottle
1.5622 to 1.5642
25g
0.9
Solubility in water: 40g/L (20°C).
OCC1=CC=CS1
114.16
114.17

Chemical Identifiers

636-72-6
114.16
ZPHGMBGIFODUMF-UHFFFAOYSA-N
69467
OCC1=CC=CS1
C5H6OS
MFCD00005454
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
(thiophen-2-yl)methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 211-264-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only