missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Thiophenemethanol, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 636-72-6
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00005454
InChI Key ZPHGMBGIFODUMF-UHFFFAOYSA-N
Synonym 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
PubChem CID 69467
IUPAC Name thiophen-2-ylmethanol
SMILES OCC1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC138860250
SDS View Documents
Thermo Scientific Chemicals
138860250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC138861000
SDS View Documents
Thermo Scientific Chemicals
138861000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

636-72-6
114.16
ZPHGMBGIFODUMF-UHFFFAOYSA-N
69467
OCC1=CC=CS1
C5H6OS
MFCD00005454
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
thiophen-2-ylmethanol

Specifications

636-72-6
100.0
206.0°C to 207.0°C (761.0mmHg)
Authentic
Glass bottle
1.5622 to 1.5642
25g
0.9
Solubility in water: 40g/L (20°C).
OCC1=CC=CS1
114.16
114.17
2-Thiophenemethanol
99.0
0.9000g/mL
>110°C
99% min. (GC)
C5H6OS
MFCD00005454
17, 113
2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
ZPHGMBGIFODUMF-UHFFFAOYSA-N
(thiophen-2-yl)methanol
69467
99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 211-264-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only