missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethoxy)phenyl Isocyanate 98.0+%, TCI America™
$121.11 - $408.30
Chemical Identifiers
CAS | 182500-26-1 |
---|---|
Molecular Formula | C8H4F3NO2 |
Molecular Weight (g/mol) | 203.12 |
MDL Number | MFCD00134160 |
InChI Key | CTMGYQHKKIEXKF-UHFFFAOYSA-N |
Synonym | 2-trifluoromethoxy phenyl isocyanate, 1-isocyanato-2-trifluoromethoxy benzene, 1-isocyanato-2-trifluoromethoxy-benzene, benzene, 1-isocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisocyanate, pubchem4460, ksc181o4h, acmc-1c053, trifluoromethoxyphenyl isocyanate, 2-trifluoromethoxyphenylisocyanate |
PubChem CID | 608953 |
IUPAC Name | 1-isocyanato-2-(trifluoromethoxy)benzene |
SMILES | C1=CC=C(C(=C1)N=C=O)OC(F)(F)F |
Chemical Identifiers
182500-26-1 | |
203.12 | |
CTMGYQHKKIEXKF-UHFFFAOYSA-N | |
608953 | |
C1=CC=C(C(=C1)N=C=O)OC(F)(F)F |
C8H4F3NO2 | |
MFCD00134160 | |
2-trifluoromethoxy phenyl isocyanate, 1-isocyanato-2-trifluoromethoxy benzene, 1-isocyanato-2-trifluoromethoxy-benzene, benzene, 1-isocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisocyanate, pubchem4460, ksc181o4h, acmc-1c053, trifluoromethoxyphenyl isocyanate, 2-trifluoromethoxyphenylisocyanate | |
1-isocyanato-2-(trifluoromethoxy)benzene |
Specifications
182500-26-1 | |
55°C | |
MFCD00134160 | |
2478 | |
CTMGYQHKKIEXKF-UHFFFAOYSA-N | |
1-isocyanato-2-(trifluoromethoxy)benzene | |
608953 | |
≥98.0% (GC) | |
2-(Trifluoromethoxy)phenyl Isocyanate |
Yellow | |
C8H4F3NO2 | |
1 g | |
2-trifluoromethoxy phenyl isocyanate, 1-isocyanato-2-trifluoromethoxy benzene, 1-isocyanato-2-trifluoromethoxy-benzene, benzene, 1-isocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisocyanate, pubchem4460, ksc181o4h, acmc-1c053, trifluoromethoxyphenyl isocyanate, 2-trifluoromethoxyphenylisocyanate | |
C1=CC=C(C(=C1)N=C=O)OC(F)(F)F | |
203.12 | |
203.12 | |
Liquid |
Safety and Handling
TSCA : No