missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Trifluoromethoxy)phenyl Isocyanate 98.0+%, TCI America™
Supplier: TCI America T29131G
Specifications
2-(Trifluoromethoxy)phenyl Isocyanate | |
Yellow | |
C8H4F3NO2 | |
1 g | |
2-trifluoromethoxy phenyl isocyanate, 1-isocyanato-2-trifluoromethoxy benzene, 1-isocyanato-2-trifluoromethoxy-benzene, benzene, 1-isocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisocyanate, pubchem4460, ksc181o4h, acmc-1c053, trifluoromethoxyphenyl isocyanate, 2-trifluoromethoxyphenylisocyanate | |
C1=CC=C(C(=C1)N=C=O)OC(F)(F)F | |
203.12 | |
203.12 | |
Liquid |
182500-26-1 | |
55°C | |
MFCD00134160 | |
2478 | |
CTMGYQHKKIEXKF-UHFFFAOYSA-N | |
1-isocyanato-2-(trifluoromethoxy)benzene | |
608953 | |
≥98.0% (GC) |
Chemical Identifiers
182500-26-1 | |
203.12 | |
CTMGYQHKKIEXKF-UHFFFAOYSA-N | |
608953 | |
C1=CC=C(C(=C1)N=C=O)OC(F)(F)F |
C8H4F3NO2 | |
MFCD00134160 | |
2-trifluoromethoxy phenyl isocyanate, 1-isocyanato-2-trifluoromethoxy benzene, 1-isocyanato-2-trifluoromethoxy-benzene, benzene, 1-isocyanato-2-trifluoromethoxy, 2-trifluoromethoxy benzenisocyanate, pubchem4460, ksc181o4h, acmc-1c053, trifluoromethoxyphenyl isocyanate, 2-trifluoromethoxyphenylisocyanate | |
1-isocyanato-2-(trifluoromethoxy)benzene |
Safety and Handling
TSCA : No